EMAIL THIS PAGE TO A FRIEND
International journal of medical sciences
Incidence and Risk Factors of Acute Kidney Injury after Radical Cystectomy: Importance of Preoperative Serum Uric Acid Level.
PMID 26283877
Abstract
Acute kidney injury (AKI) is a common complication after surgery and increases costs, morbidity, and mortality of hospitalized patients. While radical cystectomy associates significantly with an increased risk of serious complications, including AKI, risk factors of AKI after radical cystectomy has not been reported. This study was performed to determine the incidence and independent predictors of AKI after radical cystectomy. All consecutive patients who underwent radical cystectomy in 2001-2013 in a single tertiary-care center were identified. Their demographics, laboratory values, and intraoperative data were recorded. Postoperative AKI was defined and staged according to the Acute Kidney Injury Network criteria on the basis of postoperative changes in creatinine levels. Independent predictors of AKI were identified by univariate and multivariate logistic regression analyses. Of the 238 patients who met the eligibility criteria, 91 (38.2%) developed AKI. Univariate logistic regression analyses showed that male gender, high serum uric acid level, and long operation time associated with the development of AKI. On multivariate logistic regression analysis, preoperative serum uric acid concentration (odds ratio [OR] = 1.251; 95% confidence interval [CI] = 1.048-1.493; P = 0.013) and operation time (OR = 1.005; 95% CI = 1.002-1.008; P = 0.003) remained as independent predictors of AKI after radical cystectomy. AKI after radical cystectomy was a relatively common complication. Its independent risk factors were high preoperative serum uric acid concentration and long operation time. These observations can help to prevent AKI after radical cystectomy.