PloS one
Aquaporin 1 Is Involved in Acid Secretion by Ionocytes of Zebrafish Embryos through Facilitating CO2 Transport.
PMID 26287615
Abstract
Mammalian aquaporin 1 (AQP1) is well known to function as a membrane channel for H2O and CO2 transport. Zebrafish AQP1a.1 (the homologue of mammalian AQP1) was recently identified in ionocytes of embryos; however its role in ionocytes is still unclear. In this study, we hypothesized that zebrafish AQP1a.1 is involved in the acid secretion by ionocytes through facilitating H2O and CO2 diffusion. A real-time PCR showed that mRNA levels of AQP1a.1 in embryos were induced by exposure to 1% CO2 hypercapnia for 3 days. In situ hybridization and immunohistochemistry showed that the AQP1a.1 transcript was highly expressed by acid-secreting ionocytes, i.e., H+-ATPase-rich (HR) cells. A scanning ion-selective electrode technique (SIET) was applied to analyze CO2-induced H+ secretion by individual ionocytes in embryos. H+ secretion by HR cells remarkably increased after a transient loading of CO2 (1% for 10 min). AQP1a.1 knockdown with morpholino oligonucleotides decreased the H+ secretion of HR cells by about half and limited the CO2 stimulated increase. In addition, exposure to an AQP inhibitor (PCMB) for 10 min also suppressed CO2-induced H+ secretion. Results from this study support our hypothesis and provide in vivo evidence of the physiological role of AQP1 in CO2 transport.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
55540
4-(Hydroxymercuri)benzoic acid sodium salt, ≥95.0% (Hg)
C7H5HgNaO3
144207
Chlorodimethylsilane, 98%
C2H7ClSi
E10521
Ethyl 3-aminobenzoate methanesulfonate, 98%
C9H11NO2 · CH4SO3
84010
NBT Vials, for in vitro diagnostic use
C40H30Cl2N10O6
N6876
Nitrotetrazolium Blue chloride, ~98% (TLC)
C40H30Cl2N10O6
N6639
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
C40H30Cl2N10O6
P2222
Potassium phosphate dibasic, meets USP testing specifications
HK2O4P
450200
Potassium phosphate dibasic, 99.95% trace metals basis
HK2O4P
P8281
Potassium phosphate dibasic, reagent grade, ≥98.0%
HK2O4P
60353
Potassium phosphate dibasic, anhydrous, for luminescence, for molecular biology, BioUltra, ≥99.0% (T)
HK2O4P
P8584
Potassium phosphate dibasic solution, 1.0 M
HK2O4P
P5655
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
H2KO4P
RES20760-A7
Potassium phosphate monobasic, PharmaGrade, NF, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
H2KO4P
P9791
Potassium phosphate monobasic, for molecular biology, ≥98.0%
H2KO4P
229806
Potassium phosphate monobasic, 99.99% trace metals basis
H2KO4P
P5379
Potassium phosphate monobasic, ReagentPlus®
H2KO4P
60218
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
H2KO4P