EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Inactivation of the protease inhibitor phenylmethylsulfonyl fluoride in buffers.
PMID 26289
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7626
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
C7H7FO2S