EMAIL THIS PAGE TO A FRIEND
Virology
Finding balance: Virus populations reach equilibrium during the infection process.
PMID 26291064
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
559814
1-Naphthyl-N-methylcarbamate, 97%
C12H11NO2