SSEA-4 and YKL-40 positive progenitor subtypes in the subventricular zone of developing human neocortex.
PMID 26295543
The glycosphingolipid SSEA-4 and the glycoprotein YKL-40 have both been associated with human embryonic and neural stem cell differentiation. We investigated the distribution of SSEA-4 and YKL-40 positive cells in proliferative zones of human fetal forebrain using immunohistochemistry and double-labeling immunofluorescence. A few small rounded SSEA-4 and YKL-40 labeled cells were present in the radial glial BLBP positive proliferative zones adjacent to the lateral ganglionic eminence from 12th week post conception. With increasing age, a similarly stained cell population appeared more widespread in the subventricular zone. At midgestation, the entire subventricular zone showed patches of SSEA-4, YKL-40, and BLBP positive cells. Co-labeling with markers for radial glial cells (RGCs) and neuronal, glial, and microglial markers tested the lineage identity of this subpopulation of radial glial descendants. Adjacent to the ventricular zone, a minor fraction showed overlap with GFAP but not with nestin, Olig2, NG2, or S100. No co-localization was found with neuronal markers NeuN, calbindin, DCX or with markers for microglial cells (Iba-1, CD68). Moreover, the SSEA-4 and YKL-40 positive cell population in subventricular zone was largely devoid of Tbr2, a marker for intermediate neuronal progenitor cells descending from RGCs. YKL-40 has recently been found in astrocytes in the neuron-free fimbria, and both SSEA-4 and YKL-40 are present in malignant astroglial brain tumors. We suggest that the population of cells characterized by immunohistochemical combination of antibodies against SSEA-4 and YKL-40 and devoid of neuronal and microglial markers represent a yet unexplored astrogenic lineage illustrating the complexity of astroglial development.
Related Materials
Product #
Molecular Formula
Add to Cart
C7078 Casein from bovine milk, technical grade