PloS one
Isolation of Tibet orbivirus, TIBOV, from Culicoides Collected in Yunnan, China.
PMID 26295700
Abstract
We isolated a novel virus strain (YN12246) from Culicoides spp. specimens collected at the China-Laos-Myanmar border in southern Yunnan Province. This virus had a cytopathic effect (CPE) on both insect cells (C6/36) and mammalian cells (BHK-21). Electron microscopy revealed the structure of the virions to be spherical with a diameter of 75 nm. Polyacrylamide gel analysis demonstrated that the viral genome consisted of 10 segments of double-stranded RNA (dsRNA), with a distribution pattern of 3-3-3-1. The coding sequences of 9 genome segments of YN12246 (Seg1, Seg3-Seg10) were obtained by high-throughput sequencing and Sanger sequencing. Comparisons of conserved genome segments 1 and 3 (Seg1 and Seg3), encoding the polymerase-VP1 and sub-core T2 protein, respectively, showed that YN12246 groups with the Culicoides-borne orbiviruses. The highest levels of sequence identity were detected between YN12246 and Tibet orbivirus (TIBOV), indicating that they belong to the same virus species (with amino acid identity of 98.8% and 96.4% for the polymerase and T2 protein, respectively). The data presented here confirm that YN12246 is a member of the TIBOV species, which was first isolated from mosquitoes in 2009. This is the first report of the isolation of TIBOV from Culicoides.