PloS one
NaHS Protects Cochlear Hair Cells from Gentamicin-Induced Ototoxicity by Inhibiting the Mitochondrial Apoptosis Pathway.
PMID 26295804
Abstract
Aminoglycoside antibiotics such as gentamicin could cause ototoxicity in mammalians, by inducing oxidative stress and apoptosis in sensory hair cells of the cochlea. Sodium hydrosulfide (NaHS) is reported to alleviate oxidative stress and apoptosis, but its role in protecting aminoglycoside-induced hearing loss is unclear. In this study, we investigated the anti-oxidant and anti-apoptosis effect of NaHS in in vitro cultured House Ear Institute-Organ of Corti 1 (HEI-OC1) cells and isolated mouse cochlea. Results from cultured HEI-OC1 cells and cochlea consistently indicated that NaHS exhibited protective effects from gentamicin-induced ototoxicity, evident by maintained cell viability, hair cell number and cochlear morphology, reduced reactive oxygen species production and mitochondrial depolarization, as well as apoptosis activation of the intrinsic pathway. Moreover, in the isolated cochlear culture, NaHS was also demonstrated to protect the explant from gentamicin-induced mechanotransduction loss. Our study using multiple in vitro models revealed for the first time, the potential of NaHS as a therapeutic agent in protecting against aminoglycoside-induced hearing loss.