Journal of chromatography. A
A series of strategies for solving the shortage of reference standards for multi-components determination of traditional Chinese medicine, Mahoniae Caulis as a case.
PMID 26298606
In order to solve the bottleneck of reference standards shortage for comprehensive quality control of traditional Chinese medicines (TCMs), a series of strategies, including one single reference standard to determine multi-compounds (SSDMC), quantitative analysis by standardized reference extract (QASRE), and quantitative nuclear magnetic resonance spectroscopy (qNMR) were proposed, and Mahoniae Caulis was selected as an example to develop and validate these methods for simultaneous determination of four alkaloids, columbamine, jatrorrhizine, palmatine, and berberine. Comprehensive comparisons among these methods and with the conventional external standard method (ESM) were carried out. The relative expanded uncertainty of measurement was firstly used to compare their credibility. The results showed that all these three new developed methods can accurately accomplish the quantification by using only one purified reference standard, but each of them has its own advantages and disadvantages as well as the specific application scope, which were also discussed in detail in this paper.