EMAIL THIS PAGE TO A FRIEND
Virology
Blunted inflammatory and mucosal IgA responses to pneumonia virus of mice in C57BL/6 neonates are correlated to reduced protective immunity upon re-infection as elderly mice.
PMID 26298860