EMAIL THIS PAGE TO A FRIEND
Oncotarget
Whole-transcriptome analysis of flow-sorted cervical cancer samples reveals that B cell expressed TCL1A is correlated with improved survival.
PMID 26299617
Abstract
Cervical cancer is typically well infiltrated by immune cells. Because of the intricate relationship between cancer cells and immune cells, we aimed to identify both cancer cell and immune cell expressed biomarkers. Using a novel approach, we isolated RNA from flow-sorted viable EpCAM+ tumor epithelial cells and CD45+ tumor-infiltrating immune cells obtained from squamous cell cervical cancer samples (n = 24). Total RNA was sequenced and differential gene expression analysis of the CD45+ immune cell fractions identified TCL1A as a novel marker for predicting improved survival (p = 0.007). This finding was validated using qRT-PCR (p = 0.005) and partially validated using immunohistochemistry (p = 0.083). Importantly, TCL1A was found to be expressed in a subpopulation of B cells (CD3-/CD19+/CD10+/CD34-) using multicolor immunofluorescence. A high TCL1A/CD20 (B cell) ratio, determined in total tumor samples from a separate patient cohort using qRT-PCR (n = 52), was also correlated with improved survival (p = 0.027). This is the first study demonstrating the prognostic value of separating tumor epithelial cells from tumor-infiltrating immune cells and determining their RNA expression profile for identifying putative cancer biomarkers. Our results suggest that intratumoral TCL1A+ B cells are important for controlling cervical cancer development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
767638
Dinaphtho[2,3-b:2′,3′-f]thieno[3,2-b]thiophene, sublimed grade, 99%
C22H12S2