EMAIL THIS PAGE TO A FRIEND
Molecular Medicine Reports
Activation of sonic hedgehog signaling enhances cell migration and invasion by induction of matrix metalloproteinase-2 and -9 via the phosphoinositide-3 kinase/AKT signaling pathway in glioblastoma.
PMID 26299938