EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Differences in gene expression and alterations in cell cycle of acute myeloid leukemia cell lines after treatment with JAK inhibitors.
PMID 26300391
Abstract
Janus kinase (JAK) inhibitors are a promising treatment strategy in several hematological malignancies and autoimmune diseases. A number of inhibitors are in clinical development, and two have already reached the market. Unfortunately, all of them are burdened with different toxicity profiles. To check if the JAK inhibitors of different selectivity evoke different responses on JAK2-dependent and independent cells, we have used three acute myeloid leukemia cell lines with confirmed JAK2 mutation status. We have found that JAK inhibitors exert distinct effect on the expression of BCLXL, CCND1 and c-MYC genes, regulated by JAK pathway, in JAK2 wild type cells in comparison to JAK2 V617F-positive cell lines. Moreover, cell cycle analysis showed that inhibitors alter the cycle by arresting cells in different phases. Our results suggest that observed effect of JAK2 inhibitors on transcription and cell cycle level in different cell lines are associated not with activity within JAK family, but presumably with other off-target activities.