EMAIL THIS PAGE TO A FRIEND
European journal of inorganic chemistry
On the Question of Site-Selective Ligand Exchange in Carboxylate-Substituted Metal Oxo Clusters.
PMID 26300687
Abstract
Reaction of [Ti