PloS one
The Flavone Luteolin Suppresses SREBP-2 Expression and Post-Translational Activation in Hepatic Cells.
PMID 26302339
Abstract
High blood cholesterol has been associated with cardiovascular diseases. The enzyme HMG CoA reductase (HMGCR) is responsible for cholesterol synthesis, and inhibitors of this enzyme (statins) have been used clinically to control blood cholesterol. Sterol regulatory element binding protein (SREBP) -2 is a key transcription factor in cholesterol metabolism, and HMGCR is a target gene of SREBP-2. Attenuating SREBP-2 activity could potentially minimize the expression of HMGCR. Luteolin is a flavone that is commonly detected in plant foods. In the present study, Luteolin suppressed the expression of SREBP-2 at concentrations as low as 1 μM in the hepatic cell lines WRL and HepG2. This flavone also prevented the nuclear translocation of SREBP-2. Post-translational processing of SREBP-2 protein was required for nuclear translocation. Luteolin partially blocked this activation route through increased AMP kinase (AMPK) activation. At the transcriptional level, the mRNA and protein expression of SREBP-2 were reduced through luteolin. A reporter gene assay also verified that the transcription of SREBF2 was weakened in response to this flavone. The reduced expression and protein processing of SREBP-2 resulted in decreased nuclear translocation. Thus, the transcription of HMGCR was also decreased after luteolin treatment. In summary, the results of the present study showed that luteolin modulates HMGCR transcription by decreasing the expression and nuclear translocation of SREBP-2.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N5893
(±)-Naringenin, ≥95%
C15H12O5
ABS229
Anti-HMG-CoA reductase Antibody, Anti-HMG-CoA reductase Antibody detects level of HMG-CoA reductase & has been published & validated for use in Immunoprecipitation, Western Blotting.
465119
Baicalein, 98%
C15H10O5
C80105
Chrysin, 97%
C15H10O4
F2003
Flavone, ≥99%
C15H10O2
G6649
Genistein, synthetic, ≥98% (HPLC), powder
C15H10O5
G6776
Genistein, from Glycine max (soybean), ~98% (HPLC)
C15H10O5
G2911
GF 109203X, synthetic, ≥90% (HPLC)
C25H24N4O2
W431300
Hesperetin, ≥95%
C16H14O6
I3766
Isoliquiritigenin, powder
C15H12O4
P9346
Liquified Phenol, meets USP testing specifications, ≥89.0%
C6H6O
L9283
Luteolin, ≥98% (TLC), powder
C15H10O6
W530098
Naringenin, natural (US), 98%
C15H12O5
P1037
Phenol, for molecular biology
C6H6O
P5566
Phenol, BioXtra, ≥99.5% (GC)
C6H6O
328111
Phenol, unstabilized, purified by redistillation, ≥99%
C6H6O
242322
Phenol, contains hypophosphorous as stabilizer, loose crystals, ACS reagent, ≥99.0%
C6H6O
185450
Phenol, unstabilized, ReagentPlus®, ≥99%
C6H6O
77607
Phenol, BioUltra, for molecular biology, TE-saturated, ~73% (T)
C6H6O
33517
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
C6H6O
16016
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
C6H6O
16017
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
C6H6O
16018
Phenol, ≥96.0% (calc. on dry substance, T)
C6H6O
W322318
Phenol, ≥99%
C6H6O
W322340
Phenol, natural, 97%, FG
C6H6O
P4557
Phenol solution, BioReagent, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, for molecular biology
C6H6O
P4682
Phenol solution, BioReagent, Saturated with 0.1 M citrate buffer, pH 4.3 ± 0.2, for molecular biology
C6H6O
P9333
Potassium chloride, BioXtra, ≥99.0%
ClK
P9541
Potassium chloride, for molecular biology, ≥99.0%
ClK
P5405
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
ClK