EMAIL THIS PAGE TO A FRIEND
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society
Demonstration of elevated levels of active cathepsin S in dextran sulfate sodium colitis using a new activatable probe.
PMID 26303377
Abstract
Proteases play a major role in inflammatory diseases of the gastrointestinal tract. Activatable probes are a major technological advance, enabling sensitive detection of active proteases in tissue samples. Our aim was to synthesize an activatable probe for cathepsin S and validate its use in a mouse model of colitis. We designed and synthesized a new fluorescent activatable probe, NB200, for the detection of active cathepsin S. Colitis was induced in C57BL/6 mice by the administration of 3% dextran sulfate sodium (DSS). Homogenized mouse colons, with or without the addition of the specific cathepsin S inhibitor MV026031, were incubated with NB200 in a fluorescent plate reader. NB200 selectively detected purified cathepsin S and not other common inflammatory proteases. Homogenates of colon from mice with DSS colitis induced a significant fluorescent increase when compared to control animals (control vs DSS: p < 0.05 at 200 min and p < 0.01 at 220-240 min), indicating cathepsin S activation. The cathepsin S inhibitor abolished this increase in fluorescence (DSS vs DSS + MV026031: p < 0.05 at 140 min, p < 0.01 at 180 min, p < 0.001 at 200-240 min), which confirms cathepsin S activation. Cathepsin S activity correlated with the disease activity index (Spearman r = 0.77, p = 0.017). Our investigation has demonstrated the utility of activatable probes for detecting protease activity in intestinal inflammation. Panels of such probes may allow 'signature' protease profiles to be established for a range of inflammatory diseases and disorders.