EMAIL THIS PAGE TO A FRIEND
Neurochemical research
Up-Regulation of KPNB1 Involves in Neuronal Apoptosis Following Intracerebral Hemorrhage in Adult Rats.
PMID 26303509
Abstract
Kpnb1, also known as Importin β1, is a member of the Karyopherin protein family which plays a important role in nuclear import and export pathways. Its expression has been shown to be responsive to stress, such as heat shock, ethanol and oxidative stress. Previous studies demonstrated that Kpnb1 had anti-apoptotic in cervical cancer. These together prompted us to explore whether Kpnb1 has some association with neuron apoptosis in the pathophysiology of intracerebral hemorrhage (ICH). In our study, an ICH model was established by injecting into the right basal ganglia of adult rats with their autologous whole blood and assessed by behavioral tests. We found Kpnb1 were significantly up-regulated adjacent to the hematoma following ICH by Western blot and immunohistochemistry. Double immunofluorenscence manifested Kpnb1 was strikingly increased in neurons, not astrocytes or microglia. Furthermore, we also found that kpnb1 had co-localizations with active-caspase-3 which is a neuronal apoptosis marker suggesting its role in neuronal apoptosis. What's more, our in vitro study, using Kpnb1 RNA interference in PC12 cells, further indicated that Kpnb1 might exert its pro-apoptotic function on neuronal apoptosis. Therefore, Kpnb1 may play a role in the neuronal apoptosis following ICH.