EMAIL THIS PAGE TO A FRIEND
Gigiena truda i professional'nye zabolevaniia
[Experimental evaluation of the effect of aerosols of titanium mono- and disulfide on the body].
PMID 2630403
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
333492
Titanium(IV) sulfide, powder, -200 mesh, 99.9%
S2Ti