EMAIL THIS PAGE TO A FRIEND
Clinical Immunology (Orlando, Fla.)
B cells from nuclear factor kB essential modulator deficient patients fail to differentiate to antibody secreting cells in response to TLR9 ligand.
PMID 26307434