EMAIL THIS PAGE TO A FRIEND
European Journal of Mass Spectrometry (Chichester, England)
Simple mass spectrometric method for the estimation of boron and aluminum in water at the parts per billion level.
PMID 26307729