EMAIL THIS PAGE TO A FRIEND
International journal of environmental research and public health
Prevalence and Characterization of Integrons in Multidrug Resistant Acinetobacter baumannii in Eastern China: A Multiple-Hospital Study.
PMID 26308028
Abstract
The aim of this multiple-hospital study was to investigate the prevalence of integrons in multidrug-resistant Acinetobacter baumannii (MDRAB) in Eastern China, and characterize the integron-integrase genes, so as to provide evidence for the management and appropriate antibiotic use of MDRAB infections. A total of 425 clinical isolates of A. baumannii were collected from 16 tertiary hospitals in 11 cities of four provinces (Fujian, Jiangsu, Zhejiang and Shandong) from January 2009 to June 2012. The susceptibility of A. baumannii isolates to ampicillin/sulbactam, piperacillin/tazobactam, ceftazidime, ceftriaxone, cefepime, aztreonam, meropenem, amikacin, gentamicin, tobramycin, ciprofloxacin, levofloxacin, sulfamethoxazole/trimenthoprim, minocycline and imipenem was tested, and integrons and their gene cassettes were characterized in these isolates using PCR assay. In addition, integron-positive A. baumannii isolates were genotyped using pulsed-field gel electrophoresis (PFGE) assay, and intI1 gene cassette was sequenced. intI1 gene was carried in 69.6% of total A. baumannii isolates, while intI2 and intI3 genes were not detected. The prevalence of resistance to ampicillin/sulbactam, piperacillin/tazobactam, ceftazidime, ceftriaxone, cefepime, aztreonam, imipenem, meropenem, amikacin, gentamicin, tobramycin, ciprofloxacin, levofloxacin and sulfamethoxazole/trimenthoprim was significantly higher in integron-positive A. baumannii isolates than in negative isolates (all p values <0.05), while no significant difference was observed in the prevalence of minocycline resistance (p > 0.05). PFGE assay revealed 27 PFGE genotypes and 4 predominant genotypes, P1, P4, P7 and P19. The PFGE genotype P1 contained 13 extensive-drug resistant and 89 non-extensive-drug resistant A. baumannii isolates, while the genotype P4 contained 34 extensive-drug resistant and 67 non-extensive-drug resistant isolates, appearing a significant antimicrobial resistance pattern (both p values <0.05). Sequencing analysis revealed two gene cassette assays of aacA4-catB8-aadA1 and dfrXII-orfF-aadA2 in MDRAB isolates. The results of this study demonstrate a high prevalence of class 1 integrons in MDRAB in Eastern China, and a greater prevalence of antimicrobial resistance in intI1 gene-positive MDRAB isolates than in negative isolates. Four predominant PFGE genotypes are identified in intI1 gene-positive MDRAB isolates, in which P4 is an epidemic PFGE genotype in Fujian Province, and it has a high proportion of extensive drug resistant A. baumannii. The gene cassette dfrXII-orfF-aadA2 is reported, for the first time, in A. baumannii strains isolated from Fujian Province, Eastern China.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S9701
Sulbactam
C8H11NO5S