EMAIL THIS PAGE TO A FRIEND
Scientific reports
Bifunctional enhancement of oxygen reduction reaction activity on Ag catalysts due to water activation on LaMnO3 supports in alkaline media.
PMID 26310526
Abstract
Ag is considered to be one of the best candidates for oxygen reduction reaction electrocatalysts in alkaline media for application in various electrochemical energy devices. In this study, we demonstrate that water activation is a key factor in enhancing the ORR activity in alkaline media, unlike in acid environments. Ag supported on LaMnO3 having a high oxophilicity showed a markedly higher ORR activity than that on carbon with inert surfaces. Through various electrochemical tests, it was revealed that the origin of the enhanced ORR activity of Ag/LaMnO3 is the bifunctional effect mainly due to the water activation at the interface between Ag and LaMnO3. Furthermore, the ligand effect due to the charge transfer from Mn to Ag leads to the enhancement of both oxygen activation on Ag and water activation on Mn sites, and hence, an improvement in the ORR activity of Ag/LaMnO3. On the other hand, the strain effect based on the fine structure variation in the lattice was negligible. We therefore suggest that the employment of a co-catalyst or support with highly oxophilic nature and the maximization of the interface between catalyst and support should be considered in the design of electrocatalysts for the ORR in alkaline media.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S5770
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
C6H5Na3O7
83273
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
C6H5Na3O7
S8157
Silver nitrate, BioXtra, >99% (titration)
AgNO3
S7276
Silver nitrate, BioReagent, suitable for plant cell culture, >99% (titration)
AgNO3
204390
Silver nitrate, 99.9999% trace metals basis
AgNO3
792276
Silver nitrate, anhydrous, ≥99.999% trace metals basis
AgNO3
248762
Silver nitrate on silica gel, extent of labeling: ~10 wt. % loading, +230 mesh
AgNO3
85193
Silver nitrate solution, 2.5 % (w/v) AgNO3 in H2O
AgNO3
71320
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
H4BNa
452890
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
H4BNa
480886
Sodium borohydride, granular, 99.99% trace metals basis
H4BNa
452874
Sodium borohydride, granular, 10-40 mesh, 98%
H4BNa
452882
Sodium borohydride, powder, ≥98.0%
H4BNa
247677
Sodium borohydride, 90% NaBH4 basis, pellets, diam. 8 mm, Cobalt (Doped)
H4BNa
71318
Sodium borohydride, purum, ≥97.0% (gas-volumetric), tablet
H4BNa
213462
Sodium borohydride, ReagentPlus®, 99%
H4BNa
686018
Sodium borohydride, hydrogen-storage grade, 98%
H4BNa
200972
Sodium borohydride solution, 0.5 M in 2-methoxyethyl ether
H4BNa
452904
Sodium borohydride solution, SWS, ~12 wt. % in 14 M NaOH
H4BNa
215511
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
H4BNa
632287
VenPure® SF, powder
H4BNa