EMAIL THIS PAGE TO A FRIEND
SpringerPlus
Enhanced oral bioavailability of vancomycin in rats treated with long-term parenteral nutrition.
PMID 26312207
Abstract
Long-term parenteral nutrition (PN) can induce intestinal atrophy, leading to a loss of epithelial integrity in the small intestines. This change may alter the intestinal permeability of vancomycin (VCM), a non-absorbable antibiotic. The aim of the present study was to investigate the effect of PN on the pharmacokinetics of VCM in rats. VCM was intravenously (5 mg/kg) or intraduodenally (20 mg/kg) administered to control and PN rats, which were prepared by administration of PN for 9 days. After intravenous administration, there were no significant differences in any of the VCM pharmacokinetic parameters between the control and PN rats. However, after intraduodenal administration, the maximum concentration and area under the plasma concentration-time curve of VCM in PN rats was approximately 2.4- and 2.6-fold higher, respectively, than in the control rats; the calculated bioavailability was approximately 0.5 and 1.3 % in control and PN rats, respectively. These results indicated that PN administration did not affect VCM disposition, but enhanced VCM absorption; however, the enhanced oral VCM bioavailability was statistically, not clinically, significant. Therefore, while long-term PN administration may play a role in the enhancement of VCM bioavailability, this effect may be negligible without any complications.