EMAIL THIS PAGE TO A FRIEND
Ecotoxicology and Environmental Safety
Morphological alteration, lysosomal membrane fragility and apoptosis of the cells of Indian freshwater sponge exposed to washing soda (sodium carbonate).
PMID 26313128