EMAIL THIS PAGE TO A FRIEND
Revista espa?ola de cardiologia (English ed.)
Quality Markers in Cardiology. Main Markers to Measure Quality of Results (Outcomes) and Quality Measures Related to Better Results in Clinical Practice (Performance Metrics). INCARDIO (Indicadores de Calidad en Unidades Asistenciales del i??i??rea del Coraz_i??n): A SEC/SECTCV Consensus Position Paper.
PMID 26315766