EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology: Gastrointestinal and Liver Physiology
Store-operated cAMP signaling contributes to Ca2+-activated Cl- secretion in T84 colonic cells.
PMID 26316590
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P4413
N,N,N′,N′-Tetrakis(2-pyridylmethyl)ethylenediamine
C26H28N6