EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Effects of ONO-6950, a novel dual cysteinyl leukotriene 1 and 2 receptors antagonist, in a guinea pig model of asthma.
PMID 26318198
Abstract
We assessed in this study the anti-asthmatic effects of ONO-6950, a novel cysteinyl leukotriene 1 (CysLT1) and 2 (CysLT2) receptors dual antagonist, in normal and S-hexyl glutathione (S-hexyl GSH)-treated guinea pigs, and compared these effects to those of montelukast, a CysLT1 selective receptor antagonist. Treatment with S-hexyl GSH reduced animals LTC4 metabolism, allowing practical evaluation of CysLT2 receptor-mediated airway response. ONO-6950 antagonized intracellular calcium signaling via human and guinea pig CysLT1 and CysLT2 receptors with IC50 values of 1.7 and 25 nM, respectively (human receptors) and 6.3 and 8.2 nM, respectively (guinea pig receptors). In normal guinea pigs, both ONO-6950 (1 or 0.3 mg/kg, p.o.) and the CysLT1 receptor antagonist montelukast (0.3 or 0.1 mg/kg, p.o.) fully attenuated CysLT1-mediated bronchoconstriction and airway vascular hyperpermeability induced by LTD4. On the other hand, in S-hexyl GSH-treated guinea pigs ONO-6950 at 3 mg/kg, p.o. or more almost completely inhibited bronchoconstriction and airway vascular hyperpermeability elicited by LTC4, while montelukast showed only partial or negligible inhibition of these airway responses. In ovalbumin sensitized guinea pigs, treatment with S-hexyl GSH on top of pyrilamine and indomethacin rendered antigen-induced bronchoconstriction sensitive to both CysLT1 and CysLT2 receptor antagonists. ONO-6950 strongly inhibited this asthmatic response to the level attained by combination therapy with montelukast and BayCysLT2RA, a selective CysLT2 receptor antagonist. These results clearly demonstrate that ONO-6950 is an orally active dual CysLT1/LT2 receptor antagonist that may provide a novel therapeutic option for patients with asthma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5514
Pyrilamine maleate salt
C17H23N3O · C4H4O4
H6886
S-Hexylglutathione
C16H29N3O6S