EMAIL THIS PAGE TO A FRIEND
The journal of physiological sciences : JPS
Gastric acid induces mucosal H2S release in rats by upregulating mRNA and protein expression of cystathionine gamma lyase.
PMID 26319795
Abstract
It is well known that hydrogen sulfide (H2S) protects the gastric mucosa against gastric acid and other noxious stimulants by several mechanisms but until now the effect of gastric acid on H2S production has not been evaluated. This study was performed to determine the effect of basal and stimulated gastric acid secretion on mRNA and protein expression of cystathionine gamma lyase (CSE) and cystathionine beta synthase (CBS), and on mucosal release of H2S in rats. Seventy-two male rats were randomly assigned into 9 groups (8 in each)-control, distention, and pentagastrin-induced gastric acid secretion groups. The effects of 15% alcohol solution, propargylglycine (PAG), L-NAME, and pantoprazole were also investigated. Under anesthesia, animals underwent tracheostomy and midline laparotomy. A catheter was inserted into the stomach through the duodenum for gastric washout. At the end of the experiments, the animals were killed and the gastric mucosa was collected to measure H2S concentration and to quantify mRNA expression of CSE and CBS by quantitative real-time PCR, and expression of their proteins by western blot. Basal and stimulated gastric acid secretion increased mucosal levels of H2S, and mRNA and protein expression of CSE. Pantoprazole and L-NAME reversed H2S release and restored protein expression of CSE to the control level. Pantoprazole, but not propargylglycine, pretreatment inhibited the elevated level of protein expression of eNOS in response to distention-induced gastric acid secretion. Our findings indicated that NO mediated the stimulatory effect of gastric acid on H2S release and protein expression of CSE.