EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Reversed phase liquid chromatographic determination of organic acids using on-line complexation with copper(II) ion.
PMID 26320653
Abstract
We describe a simple and sensitive liquid chromatographic method for the analysis of organic acids using on-line complexation with copper(II) ion. Organic acids complexed with copper(II) ion were separated on a reversed-phase C18 column and detected by UV absorption at 240 nm. The copper(II) ion concentration in the mobile phase had a great influence on separation and sensitivity. A mobile phase consisting of 10 mM copper(II) sulfate in 5 mM sulfuric acid (pH 2.3) was used to separate nine organic acids (tartaric, malic, malonic, lactic, acetic, citric, maleic, succinic and fumaric acids). The detection limits of the examined organic acids calculated at S/N = 3 ranged from 0.6 to 100 μM. The detector signal was linear over three orders of magnitude of organic acid concentration. The method successfully measured organic acids in juice and vinegar samples.