EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Combining selection valve and mixing chamber for nanoflow gradient generation: Toward developing a liquid chromatography cartridge coupled with mass spectrometer for protein and peptide analysis.
PMID 26320807
Abstract
Toward developing a micro HPLC cartridge, we have recently built a high-pressure electroosmotic pump (EOP). However, we do not recommend people to use this pump to deliver an organic solvent directly, because it often makes the pump rate unstable. We have experimented several approaches to address this issue, but none of them are satisfactory. Here, we develop an innovative approach to address this issue. We first create an abruption (a dead-volume) within a fluid conduit. We then utilize an EOP to withdraw, via a selection valve, a train of eluent solutions having decreasing eluting power into the fluid conduit. When these solutions are further aspirated through the dead-volume, these solutions are partially mixed, smoothening concentration transitions between two adjacent eluent solutions. As these solutions are pushed back, through the dead-volume again, a smooth gradient profile is formed. In this work, we characterize this scheme for gradient formation, and we incorporate this approach with a high-pressure EOP, a nanoliter injection valve, and a capillary column, yielding a micro HPLC system. We then couple this micro HPLC with an electrospray ionization - mass spectrometer for peptide and protein separations and identifications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
279544
1-Propanol, anhydrous, 99.7%
C3H8O
W292818
1-Propanol, ≥99%, FG
C3H8O
W292826
1-Propanol, natural, ≥98%, FG
C3H8O
441090
2,2′-Azobis(2-methylpropionitrile), 98%
C8H12N4
755745
2,2′-Azobis(2-methylpropionitrile), recrystallized from methanol, 99%
C8H12N4
440159
3-(Trimethoxysilyl)propyl methacrylate, 98%
C10H20O5Si
M6514
3-(Trimethoxysilyl)propyl methacrylate, ≥97%
C10H20O5Si
A8887
Acrylamide, suitable for electrophoresis, ≥99%
C3H5NO
A3553
Acrylamide, for electrophoresis, ≥99% (HPLC), powder
C3H5NO
A9099
Acrylamide, for molecular biology, ≥99% (HPLC)
C3H5NO
A4058
Acrylamide solution, 40%, for electrophoresis, sterile-filtered
C3H5NO
A5508
Adenosine 5′-phosphosulfate sodium salt, ≥85%
C10H14N5O10PS
A3678
Ammonium persulfate, for molecular biology, for electrophoresis, ≥98%
H8N2O8S2
A7460
Ammonium persulfate, BioXtra, ≥98.0%
H8N2O8S2
09913
Ammonium persulfate, BioUltra, for molecular biology, ≥98.0% (RT)
H8N2O8S2
235865
Butyl methacrylate, 99%, contains monomethyl ether hydroquinone as inhibitor
C8H14O2
T9281
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ~99%
C6H16N2
411019
N,N,N′,N′-Tetramethylethylenediamine, ≥99.5%, purified by redistillation
C6H16N2
T22500
N,N,N′,N′-Tetramethylethylenediamine, ReagentPlus®, 99%
C6H16N2
T7024
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, for molecular biology, ≥99% (GC)
C6H16N2