EMAIL THIS PAGE TO A FRIEND
Asian-Australasian journal of animal sciences
Feeding of Dehulled-micronized Faba Bean (Vicia faba var. minor) as Substitute for Soybean Meal in Guinea Fowl Broilers: Effect on Productive Performance and Meat Quality.
PMID 26323403
Abstract
The present study aimed to assess the effect of dietary substitution of soybean meal (SBM) with dehulled-micronized faba bean (Vicia faba var. minor) in guinea fowl broilers on their growth traits, carcass quality, and meat fatty acids composition. In this trial, 120 day-old guinea fowl keets were randomly assigned to two treatments which were fed from hatch to 12 weeks of age. Birds were fed two wheat middlings-based diets comprising of a control treatment which contained SBM (78.3 g/kg) and a test diet containing dehulled-micronized faba bean (130 g/kg) as the main protein source. Substituting SBM with faba bean had no adverse effect on growth traits, dressing percentage, or breast and thigh muscles relative weight of the guinea fowls. Conversely, a decrease (p<0.05) of abdominal fat was found in guinea fowls fed the faba bean-diet. Breast muscle of birds fed faba bean had higher L* score (p<0.05) and water-holding capacity (p<0.05) than the SBM control diet. Meat from guinea fowls fed faba bean had less total lipids (p<0.05) and cholesterol (p<0.01), and higher concentrations of phospholipids (p<0.01). Feeding faba bean increased polyunsaturated fatty acid concentrations in breast meat and decreased the saturated fatty acid levels. Moreover, dietary faba bean improved the atherogenic and thrombogenic indexes in guinea fowl breast meat. Results indicated that substitution of SBM with faba bean meal in guinea fowl diet can improve carcass qualitative traits, enhancing also meat lipid profile without negatively affecting growth performance.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4515
Methyl heptadecanoate, ≥99%
C18H36O2
286087
Methyl heptadecanoate, 95%
C18H36O2