EMAIL THIS PAGE TO A FRIEND
Scientific reports
Stable magnesium peroxide at high pressure.
PMID 26323635
Abstract
Rocky planets are thought to comprise compounds of Mg and O as these are among the most abundant elements, but knowledge of their stable phases may be incomplete. MgO is known to be remarkably stable to very high pressure and chemically inert under reduced condition of the Earth's lower mantle. However, in exoplanets oxygen may be a more abundant constituent. Here, using synchrotron x-ray diffraction in laser-heated diamond anvil cells, we show that MgO and oxygen react at pressures above 96 GPa and T = 2150 K with the formation of I4/mcm MgO2. Raman spectroscopy detects the presence of a peroxide ion (O2(2-)) in the synthesized material as well as in the recovered specimen. Likewise, energy-dispersive x-ray spectroscopy confirms that the recovered sample has higher oxygen content than pure MgO. Our finding suggests that MgO2 may be present together or instead of MgO in rocky mantles and rocky planetary cores under highly oxidized conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
529699
Magnesium oxide, ≥99.99% trace metals basis
MgO
549649
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
MgO
203718
Magnesium oxide, 99.99% trace metals basis
MgO
634646
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
MgO
267317
Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis
Re
204188
Rhenium, powder, 99.995% trace metals basis
Re
267279
Rhenium, powder, −100 mesh, ≥99.9% trace metals basis
Re
459984
Rhenium, foil, thickness 1.0 mm, 99.98% trace metals basis
Re
GF57863931
Rhenium, wire reel, 0.1m, diameter 0.25mm, annealed, 99.97%
Re
GF60560513
Rhenium, wire reel, 0.1m, diameter 0.178mm, annealed, 99.97%
Re
GF90374441
Rhenium, wire reel, 0.1m, diameter 0.3mm, annealed, 99.97%
Re
GF77806367
Rhenium, wire reel, 0.1m, diameter 0.5mm, annealed, 99.97%
Re
GF63991929
Rhenium, wire reel, 0.1m, diameter 1.0mm, annealed, 99.97%
Re
GF14519156
Rhenium, wire reel, 0.2m, diameter 0.076mm, annealed, 99.97%
Re
GF59830139
Rhenium, wire reel, 0.2m, diameter 0.10mm, annealed, 99.97%
Re
GF96591875
Rhenium, wire reel, 0.2m, diameter 0.125mm, annealed, 99.97%
Re
GF02183217
Rhenium, wire reel, 0.2m, diameter 0.15mm, annealed, 99.97%
Re
GF70064658
Rhenium, wire reel, 0.2m, diameter 0.178mm, annealed, 99.97%
Re
GF32991486
Rhenium, wire reel, 0.2m, diameter 0.25mm, annealed, 99.97%
Re
GF48358435
Rhenium, wire reel, 0.2m, diameter 0.2mm, annealed, 99.97%
Re
GF96805884
Rhenium, wire reel, 0.2m, diameter 0.3mm, annealed, 99.97%
Re
GF25796010
Rhenium, wire reel, 0.2m, diameter 0.5mm, annealed, 99.97%
Re
GF98990050
Rhenium, wire reel, 0.2m, diameter 1.0mm, annealed, 99.97%
Re
GF14894163
Rhenium, wire reel, 0.5m, diameter 0.076mm, annealed, 99.97%
Re
GF78766745
Rhenium, wire reel, 0.5m, diameter 0.10mm, annealed, 99.97%
Re
GF73984797
Rhenium, wire reel, 0.5m, diameter 0.125mm, annealed, 99.97%
Re
GF72901324
Rhenium, wire reel, 0.5m, diameter 0.15mm, annealed, 99.97%
Re
GF58081945
Rhenium, wire reel, 0.5m, diameter 0.178mm, annealed, 99.97%
Re
GF23936104
Rhenium, wire reel, 0.5m, diameter 0.25mm, annealed, 99.97%
Re
GF44804325
Rhenium, wire reel, 0.5m, diameter 0.2mm, annealed, 99.97%
Re