EMAIL THIS PAGE TO A FRIEND
Autophagy
RAB24 facilitates clearance of autophagic compartments during basal conditions.
PMID 26325487
Abstract
RAB24 belongs to a family of small GTPases and has been implicated to function in autophagy. Here we confirm the intracellular localization of RAB24 to autophagic vacuoles with immuno electron microscopy and cell fractionation, and show that prenylation and guanine nucleotide binding are necessary for the targeting of RAB24 to autophagic compartments. Further, we show that RAB24 plays a role in the maturation and/or clearance of autophagic compartments under nutrient-rich conditions, but not during short amino acid starvation. Quantitative electron microscopy shows an increase in the numbers of late autophagic compartments in cells silenced for RAB24, and mRFP-GFP-LC3 probe and autophagy flux experiments indicate that this is due to a hindrance in their clearance. Formation of autophagosomes is shown to be unaffected by RAB24-silencing with siRNA. A defect in aggregate clearance in the absence of RAB24 is also shown in cells forming polyglutamine aggregates. This study places RAB24 function in the termination of the autophagic process under nutrient-rich conditions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
82320
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
C3H6O
110205
(±)-Propylene oxide, ReagentPlus®, ≥99%
C3H6O
D27802
1,4-Diazabicyclo[2.2.2]octane, ReagentPlus®, ≥99%
C6H12N2
290734
Dabco® 33-LV
C6H12N2
D6546 Dulbecco’s Modified Eagle’s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
44612 Durcupan ACM, single component B, hardener 964
G9768 Guanosine 5′-triphosphate–Agarose, aqueous glycerol suspension
I8896
IGEPAL® CA-630, for molecular biology
I6758
IPTG, ≥99% (TLC), ≤0.1% Dioxane
C9H18O5S
PHG0010
Isopropyl β-D-1-thiogalactopyranoside, PharmaGrade, Manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production.
C9H18O5S
I5502
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
C9H18O5S
I1284
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
C9H18O5S
EMU012411 MISSION® esiRNA, esiRNA targeting mouse Rab24 (esiRNA1)
D1556
OptiPrep Density Gradient Medium, used for cell and subcellular organelle isolation
P0781
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, sterile-filtered, BioReagent, suitable for cell culture
T3258
Tetracycline, ≥88.0% (HPLC)
C22H24N2O8 · xH2O
87128
Tetracycline, ≥98.0% (NT)
C22H24N2O8 · xH2O
T1426
Trypsin from bovine pancreas, TPCK Treated, essentially salt-free, lyophilized powder, ≥10,000 BAEE units/mg protein
P1379
TWEEN® 20, viscous liquid