EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim An Der Bergstrasse, Germany)
Ruthenium-Immobilized Periodic Mesoporous Organosilica: Synthesis, Characterization, and Catalytic Application for Selective Oxidation of Alkanes.
PMID 26330333