EMAIL THIS PAGE TO A FRIEND
Letters in applied microbiology
Inactivation of Burkholderia pseudomallei on environmental surfaces using spray-applied, common liquid disinfectants.
PMID 26331308
Abstract
Five commercially available liquid antimicrobials were evaluated for their ability to decontaminate common environmental surface materials, contaminated with Burkholderia pseudomallei, using a spray-based disinfectant delivery procedure. Tests were conducted at both an ambient temperature (c. 20°C) and a lower temperature (c. 12°C) condition. Nonporous materials (glass and aluminium) were more easily decontaminated than porous materials (wood, concrete and carpet). Citric acid (1%) demonstrated poor efficacy in all test conditions. Bleach (pH-adjusted), ethanol (70%), quaternary ammonium and PineSol®, demonstrated high (>6 log10 reduction) efficacies on glass and aluminium at both temperatures, but achieved varying results for wood, carpet and concrete. Temperature had minimal effect on decontamination efficacy during these tests. Much of the antimicrobial efficacy data for pathogenic micro-organisms are generated with testing that utilizes hard nonporous surface materials. These data are not directly translatable for decontaminant selection following an incident whereby complex and porous environmental surfaces are contaminated. This study presents efficacy data for spray-applied antimicrobial liquids, when used to decontaminate common environmental surfaces contaminated with Burkholderia pseudomallei. These data can help responders develop effective remediation strategies following an environmental contamination incident involving B. pseudomallei.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
518573
Aluminum, granular, <1 mm, 99.7% trace metals basis
Al
266558
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Al
433705
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Al
326852
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Al
338788
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Al
11009
Aluminum, powder, ≥91% (complexometric)
Al
326941
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Al
266574
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Al
326860
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Al
266515
Aluminum, ≤30 μm, spherical powder, ≥99%
Al
653608
Aluminum, powder, <5 μm particle size, 99.5% trace metals basis
Al
202584
Aluminum, powder, <75 μm, ≥99.95% trace metals basis
Al
266957
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Al
733369
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Al
326887
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Al
202576
Aluminum, rod, diam. × L 3.0 mm × 100 mm, 99.999% trace metals basis
Al
GF26812043
Aluminum, foam, 150x150mm, thickness 3.2mm, bulk density 0.2g/cm3, porosity 0.93, 16 pores/cm, aluminum 6101 grade, 98.5%
Al
749036
Aluminum, sputtering target, diam. × thickness 3.00 in. × 0.125 in., 99.9995% trace metals basis
Al
GF33055185
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 16 pores/cm, aluminum 6101 grade, 98.5%
Al
GF07499010
Aluminum, foam, 150x150mm, thickness 10mm, bulk density 0.2g/cm3, porosity 0.93, aluminum 6101 grade, 98.5%
Al
GF94706354
Aluminum, mesh, 150x150mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Al
GF35817217
Aluminum, sphere, 200pcs, diameter 3.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Al
GF37288020
Aluminum, rod, 1000mm, diameter 6.0mm, as drawn, 99.999%
Al
GF81923238
Aluminum, wire reel, 25m, diameter 0.25mm, as drawn, 99.5%
Al
GF98718935
Aluminum, sphere, 100pcs, diameter 1.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Al
GF86715029
Aluminum, wire reel, 10m, diameter 0.015mm, hard, 99.99%
Al
GF43921638
Aluminum, wire reel, 5m, diameter 1.5mm, as drawn, 99.999%
Al
GF10029596
Aluminum, foam, 150x150mm, thickness 6.35mm, bulk density 0.2g/cm3, porosity 0.93, 8 pores/cm, aluminum 6101 grade, 98.5%
Al
GF12712610
Aluminum, sphere, 100pcs, diameter 6.0mm, 99%
Al
GF38084430
Aluminum, wire reel, 50m, diameter 1.5mm, as drawn, 99.999%
Al