EMAIL THIS PAGE TO A FRIEND
Cancer
Antineoplastic effects of 15(S)-hydroxyeicosatetraenoic acid and 13-S-hydroxyoctadecadienoic acid in non-small cell lung cancer.
PMID 26331820
Abstract
Previous studies have shown that the levels of 15-lipoxygenase 1 (15-LOX-1) and 15-LOX-2 as well as their metabolites 13-S-hydroxyoctadecadienoic acid (13(S)-HODE) and 15(S)-hydroxyeicosatetraenoic acid (15(S)-HETE) are significantly reduced in smokers with non-small cell lung carcinoma (NSCLC). Furthermore, animal model experiments have indicated that the reduction of these molecules occurs before the establishment of cigarette smoking carcinogen-induced lung tumors, and this suggests roles in lung tumorigenesis. However, the functions of these molecules remain unknown in NSCLC. NSCLC cells were treated with exogenous 13(S)-HODE and 15(S)-HETE, and then the ways in which they affected cell function were examined. 15-LOX-1 and 15-LOX-2 were also overexpressed in tumor cells to restore these 2 enzymes to generate endogenous 13(S)-HODE and 15(S)-HETE before cell function was assessed. The application of exogenous 13(S)-HODE and 15(S)-HETE significantly enhanced the activity of peroxisome proliferator-activated receptor γ (PPARγ), inhibited cell proliferation, induced apoptosis, and activated caspases 9 and 3. The overexpression of 15-LOX-1 and 15-LOX-2 obviously promoted the endogenous levels of 13(S)-HODE and 15(S)-HETE, which were demonstrated to be more effective in the inhibition of NSCLC. This study has demonstrated that exogenous or endogenous 13(S)-HODE and 15(S)-HETE can functionally inhibit NSCLC, likely by activating PPARγ. The restoration of 15-LOX activity to increase the production of endogenous 15(S)-HETE and 13(S)-HODE may offer a novel research direction for molecular targeting treatment of smoking-related NSCLC. This strategy can potentially avoid side effects associated with the application of synthetic PPARγ ligands.