PloS one
Human Monoclonal Antibody Targeting the Heparan Sulfate Chains of Glypican-3 Inhibits HGF-Mediated Migration and Motility of Hepatocellular Carcinoma Cells.
PMID 26332121
Abstract
Heparan sulfate proteoglycans (HSPGs) participate in many processes related to tumor development, including tumorigenesis and metastasis. HSPGs contain one or more heparan sulfate (HS) chains that are covalently linked to a core protein. Glypican-3 (GPC3) is a cell surface-associated HSPG that is highly expressed in hepatocellular carcinoma (HCC). GPC3 is involved in Wnt3a-dependent HCC cell proliferation. Our previous study reported that HS20, a human monoclonal antibody targeting the HS chains on GPC3, inhibited Wnt3a/β-catenin activation. In the current study, we showed that the HS chains of GPC3 could mediate HCC cells' migration and motility. Knocking down GPC3 or targeting the HS chains by HS20 inhibited HCC cell migration and motility. However, HS20 had no effect on GPC3 knockdown cells or GPC3 negative cells. In addition, an antibody that recognizes the core protein of GPC3 did not change the rate of cell motility. HCC cell migration and motility did not respond to either canonical or non-canonical Wnt induction, but did increase under hepatocyte growth factor (HGF) treatment. HS20-treated HCC cells exhibited less ability for HGF-mediated migration and motility. Furthermore, HS20 inhibited in vitro HCC spheroid formation and liver tumor growth in mice. GPC3 interacted with HGF; however, a mutant GPC3 lacking the HS chain showed less interaction with HGF. Blocking the HS chains on GPC3 with HS20 reduced c-Met activation in HGF-treated HCC cells and 3D-cultured spheroids. Taken together, our study suggests that GPC3 is involved in HCC cell migration and motility through HS chain-mediated cooperation with the HGF/Met pathway, showing how HS targeting has potential therapeutic implications for liver cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5882
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
C5H8O2
G7651
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G7776
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G7526
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
C5H8O2
G6257
Glutaraldehyde solution, Grade II, 25% in H2O
C5H8O2
G6403
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
C5H8O2
340855
Glutaraldehyde solution, 50 wt. % in H2O
C5H8O2
49626
Glutaraldehyde solution, for electron microscopy, ~25% in H2O
C5H8O2
G4004
Glutaraldehyde solution, 25 wt. % in H2O
C5H8O2
W512303
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
C5H8O2
H1404 Hepatocyte Growth Factor human, HGF, recombinant, expressed in Baculovirus infected High-5 cells, suitable for cell culture
H9661 Hepatocyte Growth Factor human, HGF, recombinant, expressed in NSO cells, suitable for cell culture
H5791 Hepatocyte Growth Factor human, HGF, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture
SRP6014 HGF human, recombinant, expressed in CHO cells, ≥97% (SDS-PAGE), ≥97% (HPLC)
SRP6166
HGF human, recombinant, expressed in HEK 293 cells, ≥95% (SDS-PAGE)