EMAIL THIS PAGE TO A FRIEND
The Journal of Physiological Sciences : Jps
Moderate dependence of reactive oxygen species production on membrane potential in avian muscle mitochondria oxidizing glycerol 3-phosphate.
PMID 26335765
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90101
Ampliflu Red, for fluorescence, ≥98.0% (HPLC)
C14H11NO4