EMAIL THIS PAGE TO A FRIEND
BMC musculoskeletal disorders
Management of hyperlipidemia among patients with rheumatoid arthritis in the primary care setting.
PMID 26336889
Abstract
Rheumatoid arthritis (RA) has been associated with an increased risk of cardiovascular morbidity and mortality but this has not translated to optimal management of traditional cardiovascular risk factors such as hyperlipidemia. The objectives of this study were to 1) determine the prevalence of screening for hyperlipidemia in patients with RA followed by primary care practitioners (PCP); 2) examine initiation of lipid-lowering therapy in patients with an indication, and 3) assess whether proposed modifications to cardiovascular risk calculations change the percentage of RA patients with an indication for therapy. We performed a retrospective cohort study using an academic medical center-based medical record database in the United States. Patients with RA defined by the presence of at least one ICD-9 code between 2005-2010 and followed by a PCP within the health care system were included. The positive predictive value of ICD-9 codes for accurately identifying patients with RA was 96.7%. Descriptive statistics were used to report the prevalence of screening and use of lipid-lowering therapy among those with an indication. Factors associated with not receiving lipid screening were examined using logistic regression models. Indication for and receipt of therapy were then assessed before and after the application of the European Union League Against Rheumatism (EULAR) recommended multiplier to the Framingham risk score. Among 1,056 patients with RA followed by PCPs and eligible for lipid screening, lipid screening was ordered for 539 (51%) within the 3-year follow-up period. Patients with diabetes, hypertension, chronic kidney disease, obesity or age >50 were more likely to be screened. Of those with lipid results (N = 290), 25 (9%) patients had an indication for lipid-lowering therapy based on Adult Treatment Panel III guidelines. Ten (40%) patients with an indication for lipid-lowering therapy received therapy did not receive therapy. Applying the EULAR multiplier only changed the indication for lipid-lowering therapy in two patients. Screening and management of traditional cardiovascular risk factors, including hyperlipidemia, need to be optimized.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6020
Fenofibrate, ≥99%, powder
C20H21ClO4
G9518
Gemfibrozil
C15H22O3