EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology: Gastrointestinal and Liver Physiology
The p66Shc protein controls redox signaling and oxidation-dependent DNA damage in human liver cells.
PMID 26336926