EMAIL THIS PAGE TO A FRIEND
Virology
Comprehensive mapping of functional epitopes on dengue virus glycoprotein E DIII for binding to broadly neutralizing antibodies 4E11 and 4E5A by phage display.
PMID 26339794