EMAIL THIS PAGE TO A FRIEND
European Journal of Endocrinology
Hormonal circadian rhythms in patients with congenital adrenal hyperplasia: identifying optimal monitoring times and novel disease biomarkers.
PMID 26340969