EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology. Lung Cellular and Molecular Physiology
Novel identification of the free fatty acid receptor FFAR1 that promotes contraction in airway smooth muscle.
PMID 26342087
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G9797
GW9508, ≥98% (HPLC)
C22H21NO3