EMAIL THIS PAGE TO A FRIEND
The Journal of Pain : Official Journal of the American Pain Society
Contribution of Endogenous Spinal Endomorphin 2 to Intrathecal Opioid Antinociception in Rats Is Agonist Dependent and Sexually Dimorphic.
PMID 26342648
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
301841
1,7-Phenanthroline, 99%
C12H8N2
F9397
Flutamide
C11H11F3N2O3