EMAIL THIS PAGE TO A FRIEND
Clinica Chimica Acta; International Journal of Clinical Chemistry
Aptamer-based microchip electrophoresis assays for amplification detection of carcinoembryonic antigen.
PMID 26344338