EMAIL THIS PAGE TO A FRIEND
Clinical interventions in aging
Significant association between parathyroid hormone and uric acid level in men.
PMID 26346636
Abstract
Previous reports of patients undergoing parathyroidectomy and of patients receiving teriparatide as antiosteoporotic treatment have suggested a plausible relationship between parathyroid hormone (PTH) and uric acid. However, similar data at population level were lacking. The current study aimed to determine the relationship between PTH and uric acid in a group of apparently healthy Malaysian men. A cross-sectional study was conducted among 380 Malay and Chinese men aged 20 years and above, residing in the Klang Valley, Malaysia. Their body anthropometry was measured, and their fasting blood samples were collected for biochemical analysis. The relationship between PTH and uric acid was analyzed using regression analysis. Increased serum PTH level was significantly associated with increased serum uric acid level (β=0.165; P=0.001). Increased PTH level was also significantly associated with the condition of hyperuricemia in the study population (odds ratio [OR], 1.045; 95% confidence interval [CI], 1.017-1.075; P=0.002). All analyses were adjusted for age, body mass index, vitamin D, total calcium, inorganic phosphate, blood urea nitrogen and creatinine levels. There is a significant positive relationship between PTH level and uric acid level in Malaysian men. This relationship and its clinical significance should be further investigated in a larger longitudinal study.
Related Materials