EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
A simple synthesis of nitrogen doped porous graphitic carbon: Electrochemical determination of paracetamol in presence of ascorbic acid and p-aminophenol.
PMID 26347171
Abstract
Graphite paste electrode modified with nitrogen doped porous carbon (NDPC) is used for the detections of paracetamol (PCM), ascorbic acid (AA) and p-aminophenol (PAP) at relatively low concentration. NDPC is synthesized by direct carbonization of Zn(OAc)2 incorporated melamine-formaldehyde resin microsphere. The NDPC shows small pore diameters centered at 3.14 nm and 8.12 nm and has a pseudo graphitic structure with reasonable porous matrix. The lower limit of detections (S/N = 3) for PCM, AA, and PAP are found to be 30 nM, 720 nM and 10 nM respectively. Under optimized experimental condition, the linear ranges of determination for PCM and AA are 1-400 μM, 10-2700 μM respectively in mixture. Similarly for PCM and PAP mixture, the linear ranges of determination are found to be 1-90 μM. It is also used for the analysis of urine and pharmaceutical products with better sensitivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
47608
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
CH2O
A5960
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
C6H8O6
A7506
L-Ascorbic acid, reagent grade, crystalline
C6H8O6
A0278
L-Ascorbic acid, reagent grade
C6H8O6
A4403
L-Ascorbic acid, powder, cell culture tested, γ-irradiated
C6H8O6
A4544
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
C6H8O6
A2218
L-Ascorbic acid, meets USP testing specifications
C6H8O6
A2174
L-Ascorbic acid, plant cell culture tested
C6H8O6
95209
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
C6H8O6
33034
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.7-100.5% (oxidimetric)
C6H8O6
95210
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
C6H8O6
255564
L-Ascorbic acid, ACS reagent, ≥99%
C6H8O6
A92902
L-Ascorbic acid, 99%
C6H8O6
W210901
L-Ascorbic acid, FCC, FG
C6H8O6
M2659
Melamine, 99%
C3H6N6
295574
Nitrogen, ≥99.998%
N2
P0838
Potassium bromide, BioXtra, ≥99.0%
BrK
449962
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
BrK
451010
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
BrK
449970
Potassium bromide, anhydrous, beads, −10 mesh, ≥99.9% trace metals basis
BrK
229792
Potassium bromide, 99.99% trace metals basis
BrK
60089
Potassium bromide, BioUltra, ≥99.5% (AT)
BrK
RDD007
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
H2NaO4P
S8282
Sodium phosphate monobasic, BioXtra, ≥99.0%
H2NaO4P
S3139
Sodium phosphate monobasic, BioReagent, for molecular biology, anhydrous, ≥98%
H2NaO4P
S5011
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
H2NaO4P
S2554
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
H2NaO4P
229903
Sodium phosphate monobasic, 99.999% trace metals basis
H2NaO4P
S0751
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
H2NaO4P
74092
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
H2NaO4P