EMAIL THIS PAGE TO A FRIEND
Scientific reports
Secreted tyrosine sulfated-eIF5A mediates oxidative stress-induced apoptosis.
PMID 26348594
Abstract
Oxidative stress plays a critical role in ischemia/reperfusion-injury, atherosclerosis, and aging. It causes cell damage that leads to apoptosis via uncertain mechanisms. Because conditioned medium from cardiac myocytes subjected to hypoxia/reoxygenation induces extensive apoptosis of cardiac myocytes under normoxia, we hypothesized that a humoral factor released from the hypoxic/reoxygenated cardiac myocytes mediates apoptosis. We identified an apoptosis-inducing humoral factor in the hypoxia/reoxygenation-conditioned medium. Here, we found that eIF5A undergoes tyrosine sulfation in the trans-Golgi and is rapidly secreted from cardiac myocytes in response to hypoxia/reoxygenation; then, eIF5A induces apoptosis by acting as a pro-apoptotic ligand. The apoptosis of cardiac myocytes induced by hypoxia/reoxygenation or ultraviolet irradiation was suppressed by anti-eIF5A neutralizing monoclonal antibodies (mAbs) in vitro. Myocardial ischemia/reperfusion (but not ischemia alone) markedly increased the plasma levels of eIF5A, and treatment with anti-eIF5A neutralizing mAbs significantly reduced myocardial injury. These results identify an important, novel specific biomarker and a critical therapeutic target for oxidative stress-induced cell injury.