EMAIL THIS PAGE TO A FRIEND
Scandinavian Journal of Work, Environment & Health
Dysregulation of markers of oxidative stress and DNA damage among nail technicians despite low exposure to volatile organic compounds.
PMID 26348975