EMAIL THIS PAGE TO A FRIEND
Journal of Bacteriology
Physiological Roles and Adverse Effects of the Two Cystine Importers of Escherichia coli.
PMID 26350134
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8630
DL-Cystine
C6H12N2O4S2