EMAIL THIS PAGE TO A FRIEND
Child's nervous system : ChNS : official journal of the International Society for Pediatric Neurosurgery
Intraoperative neurophysiology in posterior fossa tumor surgery in children.
PMID 26351231